ContactUsLinkComponent
Monensin Solution (1000X), protein transport inhibitor that can be added to cell culture medium during in vitro activation of cells to inhibit the secretion of cytokines, Addition of Monensin leads to an accumulation of newly synthesized protein, Application: Functional Assays, Size: 1ml
Catalog #: 76447-260
Supplier:  Tonbo Biosciences
Monensin Solution (1000X), protein transport inhibitor that can be added to cell culture medium during in vitro activation of cells to inhibit the secretion of cytokines, Addition of Monensin leads to an accumulation of newly synthesized protein, Application: Functional Assays, Size: 1ml
Catalog #: 76447-260
Supplier:  Tonbo Biosciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.