ContactUsLinkComponent
MHC Class II monoclonal Antibody, Clone: M5/114.15.2, Host: Rat, Species reactivity: Mouse, Isotype: IgG2b, K, Format: violetFluor 500, Synonyms: MHC II, IA, IE, I-A/E, IA/IE, Applications: Flow Cytometryreacts with mouse MHC Class II alloantigens I-Ab, I-Ad, I-Aq, I-Ed, and I-Ek, size: 25ug
Catalog #: 76447-236
Supplier:  Tonbo Biosciences
MHC Class II monoclonal Antibody, Clone: M5/114.15.2, Host: Rat, Species reactivity: Mouse, Isotype: IgG2b, K, Format: violetFluor 500, Synonyms: MHC II, IA, IE, I-A/E, IA/IE, Applications: Flow Cytometryreacts with mouse MHC Class II alloantigens I-Ab, I-Ad, I-Aq, I-Ed, and I-Ek, size: 25ug
Catalog #: 76447-236
Supplier:  Tonbo Biosciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.