ContactUsLinkComponent
HLA-DR monoclonal Antibody, Clone: LN3, Host: Mouse, Species reactivity: Human, Isotype: IgG2b, Format: violetFluor 450, Applications: Flow Cytometryreacts with a member of the human MHC Class II antigens, HLA-DR, antigen is expressed on human antigen presenting cells, size: 25 tests
Catalog #: 76447-204
Supplier:  Tonbo Biosciences
HLA-DR monoclonal Antibody, Clone: LN3, Host: Mouse, Species reactivity: Human, Isotype: IgG2b, Format: violetFluor 450, Applications: Flow Cytometryreacts with a member of the human MHC Class II antigens, HLA-DR, antigen is expressed on human antigen presenting cells, size: 25 tests
Catalog #: 76447-204
Supplier:  Tonbo Biosciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.