ContactUsLinkComponent
CD24 monoclonal Antibody, Clone: M1/69, Host: Rat, Species reactivity: Mouse, Isotype: IgG2b, K, Format: PerCP-Cyanine5.5, Synonyms: Heat Stable Antigen, HSA, Applications: Flow Cytometryreacts with mouse CD24, a cell surface molecule also known as Heat Stable Antigen (HSA), size: 25ug
Catalog #: 76447-200
Supplier:  Tonbo Biosciences
CD24 monoclonal Antibody, Clone: M1/69, Host: Rat, Species reactivity: Mouse, Isotype: IgG2b, K, Format: PerCP-Cyanine5.5, Synonyms: Heat Stable Antigen, HSA, Applications: Flow Cytometryreacts with mouse CD24, a cell surface molecule also known as Heat Stable Antigen (HSA), size: 25ug
Catalog #: 76447-200
Supplier:  Tonbo Biosciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.