Quick Sars-Cov-2 rrt-pcr Kit, High Sensitivity: Limit of Detection as low as 15 copies, Rapid and Easy Setup: Ready-to-use Master Mix, just add sample, Compatible with Automated and High-Throughput workflows, Approved by U.S. Food and Drug Administration/CE-IVD certified, Size: 1000 reactions
Catalog #: 76446-942
Supplier: Zymo Research
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.