ContactUsLinkComponent
Volkman Bone Curette, 0 number, Oval Tip, Tip Configuration: Oval, Curvature: Straight, Working Surface Style: Sharp/Blunt, Material: Stainless Steel, Reusable, Non-Sterile, Latex-Free, Grade: Premium OR-Grade, Size: 6-3/4 in length, 5.2 mm
Catalog #: 76446-916
Supplier:  Sklar
Volkman Bone Curette, 0 number, Oval Tip, Tip Configuration: Oval, Curvature: Straight, Working Surface Style: Sharp/Blunt, Material: Stainless Steel, Reusable, Non-Sterile, Latex-Free, Grade: Premium OR-Grade, Size: 6-3/4 in length, 5.2 mm
Catalog #: 76446-916
Supplier:  Sklar
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.