DNA/RNA Shield Saliva Collection Kit, DX are specimen receptacle medical devices for molecular-based in vitro diagnostic applications, This DNA and RNA saliva collection kit also takes a microbial snapshot of a sample while inactivating viruses making samples safe and ready for transport, Size: 2ml
Catalog #: 76443-118
Supplier: Zymo Research
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.