ContactUsLinkComponent
DNA/RNA Shield Collection Tube, DNA/RNA Shield Collection 12 x 80 mm screwcap vial pre-filled with DNA/RNA Shield, For molecular-based in vitro diagnostic applications. Many sample types can be collected with these swab collection tubes including from the mouth, nose, throat, etc, Size: 2 ml
Catalog #: 76443-116
Supplier:  Zymo Research
DNA/RNA Shield Collection Tube, DNA/RNA Shield Collection 12 x 80 mm screwcap vial pre-filled with DNA/RNA Shield, For molecular-based in vitro diagnostic applications. Many sample types can be collected with these swab collection tubes including from the mouth, nose, throat, etc, Size: 2 ml
Catalog #: 76443-116
Supplier:  Zymo Research
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.