ContactUsLinkComponent
Safety Glasses, SR3, Clear Anti-Fog Lenses, Aqua Blue Temples, Dielectric, no metal, Lightweight and balanced frame with zero removable parts, Rugged polycarbonate lenses filters 99.9% of harmful U.V. Rays, Meets or exceeds ANSI Z87+ high impact standard
Catalog #: 76446-756
Supplier:  MCR Safety
Safety Glasses, SR3, Clear Anti-Fog Lenses, Aqua Blue Temples, Dielectric, no metal, Lightweight and balanced frame with zero removable parts, Rugged polycarbonate lenses filters 99.9% of harmful U.V. Rays, Meets or exceeds ANSI Z87+ high impact standard
Safety Glasses, SR3, Clear Anti-Fog Lenses, Aqua Blue Temples, Dielectric, no metal, Lightweight and balanced frame with zero removable parts, Rugged polycarbonate lenses filters 99.9% of harmful U.V. Rays, Meets or exceeds ANSI Z87+ high impact standard
Catalog #: 76446-756
Supplier:  MCR Safety
Customers Who Bought This Also Bought