VWR® Disposable Face Shields
Catalog #: 76443-134
Supplier: VWR International
Specifications
- Description:Disposable face shields
- Thickness:0.25 mm
Specifications
About this item
Perfect for use in environments with light particle, splash, or spray, the VWR® Disposable Face Shield is a cost effective and reliable way to keep covered.
- Material: APET
- Anti-fog inner and outer coating
- PE protective film
- Comfortable PU foam for extended wear
- Elastic band to maintain comfortable fit
Face shields should be worn with ANSI approved respiratory protection, as needed.
Packaging: Packaged 200 per case.
Customers Who Bought This Also Bought