Holey Thick Carbon Grid, Copper only, ultra-flat, for TEM, Manufactured without plastics, clean and without residue, Ultra-flat for better particle dispersion and more uniform ice thickness, Multihole device has staggered pattern of six features, Hole size: 4.0 um, Hole spacing: 2.0um, TEM mesh: 300
Catalog #: 76439-692
Supplier: ELECTRON MICROSCOPY SCIENCE
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.