Holey Thick Carbon Grid, Copper only, An ultra-flat, for TEM, Manufactured without plastics, it is clean and without residue, Ultra-flat for better particle dispersion and more uniform ice thickness, Multihole device has a staggered pattern of six features, TEM mesh: 300
Catalog #: 76439-690
Supplier: ELECTRON MICROSCOPY SCIENCE
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.