Holey Thick Carbon Grid, Copper Only, An ultra-flat, for TEM, Manufactured without plastics, it is clean and without residue, Ultra-flat for better particle dispersion and more uniform ice thickness, Hole Size: 2um, Hole spacing: 4 um, TEM Mesh: 300
Catalog #: 76439-654
Supplier: ELECTRON MICROSCOPY SCIENCE
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.