Magic wand Fine tip/Flat strong tip, Material: pvdf, Made from PEEK high performance plastic with chemical resistantance, Withstands temperatures to 300 degree C, For electronic areas to labss, from preparation of samples to applying adhesives, Use as a stirring rod or dosing samples.
Catalog #: 76439-462
Supplier: ELECTRON MICROSCOPY SCIENCE
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.