ContactUsLinkComponent
Kit, Electronic Rechargeable Decapper Includes charger with Universal Plug Set, with OLED display screen, Hundreds of operations from one charge, Large battery allows for faster charge cycle and two sleep mode options extend battery life, Certification/Compliance:CE, RoHS, REACH, Size:20 mm
Catalog #: 76437-500
Supplier:  Restek
Kit, Electronic Rechargeable Decapper Includes charger with Universal Plug Set, with OLED display screen, Hundreds of operations from one charge, Large battery allows for faster charge cycle and two sleep mode options extend battery life, Certification/Compliance:CE, RoHS, REACH, Size:20 mm
Catalog #: 76437-500
Supplier:  Restek
Customers Who Bought This Also Bought