ContactUsLinkComponent
Crimping Tool, High-Power Electronic, with OLED Display Screen, power cable for Taiwan, Certification/Compliance:CE, RoHS, REACH, No batteries to change-fixed power supply, Interchangeable jaw sets, Strong enough for steel and magnetic caps, Strong enough for steel and magnetic caps.
Catalog #: 76437-488
Supplier:  Restek
Crimping Tool, High-Power Electronic, with OLED Display Screen, power cable for Taiwan, Certification/Compliance:CE, RoHS, REACH, No batteries to change-fixed power supply, Interchangeable jaw sets, Strong enough for steel and magnetic caps, Strong enough for steel and magnetic caps.
Catalog #: 76437-488
Supplier:  Restek
Customers Who Bought This Also Bought