Inlet Liner, Geometry: mini-Lam, ID:4.0 mm, Length:78.5 mm, OD:6.3 mm, Deactivation:Standard, Material:Borosilicate Glass, for TRACE 1300/1310 GCs equipped with SSL inlets
Catalog #: 76437-482
Supplier: Restek
Inlet Liner, Geometry: mini-Lam, ID:4.0 mm, Length:78.5 mm, OD:6.3 mm, Deactivation:Standard, Material:Borosilicate Glass, for TRACE 1300/1310 GCs equipped with SSL inlets
Catalog #: 76437-482
Supplier: Restek
Customers Who Bought This Also Bought
VWR® Screw-Thread Vials 8-425, Caps and Inserts with Standard Opening
Supplier: VWR International
$64.11 to $458.29
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Supplier: VWR International
$118.61 to $1,128.11
VWR® 11 mm Crimp Top Vials, Kits, and Inserts, Standard Opening
Supplier: VWR International
$62.00 to $438.00
Advanced Green Septa for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
$165.70
Closures for 2 ml Screw Top Vials, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
$130.00
SureSTART™ Glass Inserts for 2 ml Vials, Level 3 High Performance Applications, Thermo Scientific
Supplier: Thermo Scientific
$93.60 to $97.09
WHEATON® E-Z Vial® Large-Opening Crimp-Top Vials, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
$125.61
Closures for 2 ml Crimp Top Vials, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
$28.24