Socorex® Qualitix® Sterilization Pouches, DWK Life Sciences
Catalog #: 76437-384
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Dimensions:300×395 mm
- Description:Autoclave pouches
- Packaging:Case of 200
Specifications
About this item
The Qualitix® pouches provide easy and effective wrapping of items requiring sterilization.
- Robust durable material
- Large self-sealing strip, easy to close
- Chevron top seal for easy opening
- Suitable for steam and EO gas sterilization
- Two sterilization indicators
- Five different sizes
Fitting any small to mid-size laboratory device, the Qualitix® single use pouches provide easy and effective wrapping of items requiring sterilization. Made with robust medical grade paper, the pouches tightly close with a self-sealing strip to maintain sterility. Content remains always visible through PET / CPP transparent laminated film.
Certifications: Conformity to ISO 17665-06, ISO 11135, CE.
Customers Who Bought This Also Bought