Empty Racks for Pipette Tips, Socorex® Qualitix® Premium, DWK Life Sciences
Catalog #: 76437-332
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Description:Empty tip racks
- Cat. no.:76437-332
- For Use With:10/200 µl tips
Specifications
About this item
Empty racks for tip refilling.
- Superior quality, autoclavable
- Lot by lot consistency
- Certified free of nuclease, endotoxin and ATP
- Reduced packaging size, less bench space requirement
- Broad compatibility with major micropipette brands
Customers Who Bought This Also Bought