ContactUsLinkComponent
Polymeric SPE Cartridge, WAX, 6ml/500mg, (Reversed Phase), Analyte: Strong acids, Silica-free, bonded polymeric material, High surface area, Stable over a wide pH range (0 -14), Water-wettable, No flow-rate dependence, maintains retention, even if dried out from vacuum or positive pressure flows
Catalog #: 76437-304
Supplier:  Restek
Polymeric SPE Cartridge, WAX, 6ml/500mg, (Reversed Phase), Analyte: Strong acids, Silica-free, bonded polymeric material, High surface area, Stable over a wide pH range (0 -14), Water-wettable, No flow-rate dependence, maintains retention, even if dried out from vacuum or positive pressure flows
Catalog #: 76437-304
Supplier:  Restek
Customers Who Bought This Also Bought