ContactUsLinkComponent
Polymeric SPE 96-Well Plate, MAX, 10mg, Analyte: Acids, Silica-free, bonded polymeric material, High surface area, Stable over a wide pH range (0 -14), Water-wettable, No flow-rate dependence-maintains retention and capacity after conditioning, for high throughput and automation
Catalog #: 76437-282
Supplier:  Restek
Polymeric SPE 96-Well Plate, MAX, 10mg, Analyte: Acids, Silica-free, bonded polymeric material, High surface area, Stable over a wide pH range (0 -14), Water-wettable, No flow-rate dependence-maintains retention and capacity after conditioning, for high throughput and automation
Catalog #: 76437-282
Supplier:  Restek
Customers Who Bought This Also Bought