ContactUsLinkComponent
Polymeric SPE Cartridge, MAX, 6ml/150mg, (Reversed Phase), Analyte: Acids, Silica-free, bonded polymeric material, High surface area, Stable over a wide pH range (0 -14), Water-wettable, No flow-rate dependence, maintains retention, even if dried out from vacuum or positive pressure flows
Catalog #: 76437-278
Supplier:  Restek
Polymeric SPE Cartridge, MAX, 6ml/150mg, (Reversed Phase), Analyte: Acids, Silica-free, bonded polymeric material, High surface area, Stable over a wide pH range (0 -14), Water-wettable, No flow-rate dependence, maintains retention, even if dried out from vacuum or positive pressure flows
Catalog #: 76437-278
Supplier:  Restek
Customers Who Bought This Also Bought