ContactUsLinkComponent
Polymeric SPE 96-Well Plate, HLB, 10mg, Analyte: General-purpose for acids, bases, or neutrals, Silica-free, bonded polymeric material, High surface area, Stable over a wide pH range (0 -14), Water-wettable, No flow-rate dependence-maintains retention and capacity after conditioning
Catalog #: 76437-270
Supplier:  Restek
Polymeric SPE 96-Well Plate, HLB, 10mg, Analyte: General-purpose for acids, bases, or neutrals, Silica-free, bonded polymeric material, High surface area, Stable over a wide pH range (0 -14), Water-wettable, No flow-rate dependence-maintains retention and capacity after conditioning
Catalog #: 76437-270
Supplier:  Restek
Customers Who Bought This Also Bought