Polymeric SPE Cartridge, HLB, 6ml/200mg, (Reversed Phase), Analyte: General-purpose for acids, bases, or neutrals; high capacity for polar compounds, Silica-free, bonded polymeric material, High surface area, Stable over a wide pH range (0 -14), Water-wettable, No flow-rate dependence
Catalog #: 76437-266
Supplier: Restek
Customers Who Bought This Also Bought