Solid-Top Cap, Polyethylene, Color:Clear, Compatible with all 2.0 mL, 9 mm screw-thread vials, Molded, 10-mil, solid pierceable cap
Catalog #: 76437-256
Supplier: Restek
Solid-Top Cap, Polyethylene, Color:Clear, Compatible with all 2.0 mL, 9 mm screw-thread vials, Molded, 10-mil, solid pierceable cap
Catalog #: 76437-256
Supplier: Restek
Customers Who Bought This Also Bought
WEBCOL® Alcohol Preps, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
15,59 $ to 18,12 $
Reagent Reservoirs, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
277,83 $ to 277,83 $