Capillary Column, Rxi-65TG, Length:25m, 0.32mm ID, df: 0.10um, Temp. Limits:40 to 370 deg C, (fused silica), High-polarity Crossbond phase, Ideal for the analysis of triglycerides in edible oils, Separate and quantify critical triglycerides (acylglycerols) without interference from column bleed
Catalog #: 76437-232
Supplier: Restek
Capillary Column, Rxi-65TG, Length:25m, 0.32mm ID, df: 0.10um, Temp. Limits:40 to 370 deg C, (fused silica), High-polarity Crossbond phase, Ideal for the analysis of triglycerides in edible oils, Separate and quantify critical triglycerides (acylglycerols) without interference from column bleed
Catalog #: 76437-232
Supplier: Restek
Customers Who Bought This Also Bought
SureSTART™ Caps for 2 ml Glass Snap Top Vials, Level 2 High-Throughput Applications, Thermo Scientific
Supplier: Thermo Scientific
$1,684.96 to $1,684.96
Screw Top Vials and Caps, 2 ml, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
$339.00
CrossLab Flame Ionization Detector (FID) for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
$144.30
WHEATON® ABC Vial™ Large-Opening Screw-Thread Vials, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
$131.04
RSA™ and RSA-Pro™ Autosampler Vials, Caps and Convenient Kits
Supplier: MicroSolv Technology Corp
$433.35
SureSTART™ Septa for 20 mm Stoppers, Level 2 High-throughput Applications, Thermo Scientific
Supplier: Thermo Scientific
$131.41 to $136.31
SureSTART™ Caps for 20 ml Glass Crimp Top Headspace Vials, Level 1 Everyday Analysis, Thermo Scientific
Supplier: Thermo Scientific
$1,418.93 to $1,418.93
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Supplier: VWR International
$149.40
WHEATON® MicroLiter µLPlate® Component Kits, Inserts and Caps, Unassembled, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
$180.74
SureSTART™ Screw Caps for Glass Screw Top Vials, 9 mm, Level 1 Everyday Analysis, Thermo Scientific
Supplier: Thermo Scientific
$2,106.19 to $2,106.19