ContactUsLinkComponent
Capillary Column, Rxi-65TG, Length:30m, 0.32mm ID, df: 0.10um, Temp. Limits:40 to 370 deg C, (fused silica), High-polarity Crossbond phase, Ideal for the analysis of triglycerides in edible oils, Separate and quantify critical triglycerides (acylglycerols) without interference from column bleed
Catalog #: 76437-230
Supplier:  Restek
Capillary Column, Rxi-65TG, Length:30m, 0.32mm ID, df: 0.10um, Temp. Limits:40 to 370 deg C, (fused silica), High-polarity Crossbond phase, Ideal for the analysis of triglycerides in edible oils, Separate and quantify critical triglycerides (acylglycerols) without interference from column bleed
Catalog #: 76437-230
Supplier:  Restek
Customers Who Bought This Also Bought