ContactUsLinkComponent
Allure Acrylamide LC Column, USP L3, Particle Size: 5um, Pore Size: 60A, Stable retention times for hundreds of matrix injections, Faster equilibration times allow for method optimization and shorter analyses, Accurate low ppt-level limits for acrylamide in drinking water, Dimension: 150x3.0mm
Catalog #: 76436-432
Supplier:  Restek
Allure Acrylamide LC Column, USP L3, Particle Size: 5um, Pore Size: 60A, Stable retention times for hundreds of matrix injections, Faster equilibration times allow for method optimization and shorter analyses, Accurate low ppt-level limits for acrylamide in drinking water, Dimension: 150x3.0mm
Catalog #: 76436-432
Supplier:  Restek
Customers Who Bought This Also Bought