Alert Software license required to send an unlimited number of text and voice call alerts, License based on the number of sensors, Intuitive dashboard simplifies daily usage, Optional floor plan and sites views, Remote data access, License validity: 12 months after activation
Catalog #: 76436-386
Supplier: Thermo Fisher Scientific
Customers Who Bought This Also Bought