Ecomparator, EC 2000 Gardner, Progressing from Visual to Automatic Colour Measurement, Colour Type: Transparent, Display: Size: 3.5in; Resolution: 320x240; Colour: 24Bit, Interface: USB, Keypad: 3 key tactile membrane, Instrument Housing: Flame Retardant ABS, Portable, Dimensions: 106x57x210mm
Catalog #: 76424-360
Supplier: Lovibond
Customers Who Bought This Also Bought