Ecomparator, EC 2000 Pt-Co, Colour measurement of water, Colour Type: Transparent, Display: Size: 3.5 inch; Resolution: 320 x 240; Colour: 24 Bit, Interface: USB, Keypad: 3 key tactile membrane, Instrument Housing: Flame Retardant ABS, without touchscreen, Portable, Dimensions: 106 x 57 x 210 mm
Catalog #: 76424-358
Supplier: Lovibond
Customers Who Bought This Also Bought