ContactUsLinkComponent
Gloss Meter, TG 60, Entry-Level Semi Gloss Measurement at 60 Angle, Colour Type: Opaque, Display: TFT 3.5 inch, capacitive touch screen, Interfaces: USB, RS 232, Measuring Range: 0 - 300 GU, Power Supply: 3200 mAh Li-ion Battery, Portable, Storage Temperature -20-50 C, Dimensions: 160 x 90 x 75 mm
Catalog #: 76424-356
Supplier:  Lovibond
Gloss Meter, TG 60, Entry-Level Semi Gloss Measurement at 60 Angle, Colour Type: Opaque, Display: TFT 3.5 inch, capacitive touch screen, Interfaces: USB, RS 232, Measuring Range: 0 - 300 GU, Power Supply: 3200 mAh Li-ion Battery, Portable, Storage Temperature -20-50 C, Dimensions: 160 x 90 x 75 mm
Gloss Meter, TG 60, Entry-Level Semi Gloss Measurement at 60 Angle, Colour Type: Opaque, Display: TFT 3.5 inch, capacitive touch screen, Interfaces: USB, RS 232, Measuring Range: 0 - 300 GU, Power Supply: 3200 mAh Li-ion Battery, Portable, Storage Temperature -20-50 C, Dimensions: 160 x 90 x 75 mm
Catalog #: 76424-356
Supplier:  Lovibond
Customers Who Bought This Also Bought