Order Entry
ContactUsLinkComponent
BCYE Agar/BCYE Agar without Cysteine, Optically clear polystyrene, Heavy-Duty, Stacking rings, Breathable Packaging, recommended for use in the cultivation and primary isolation of Legionella spp. In water and other samples suspected of harboring the bacteria, Dimension: 15x100mm biplate
BCYE Agar/BCYE without Cysteine, Hardy Diagnostics
Catalog #: 76436-452
Supplier:  Hardy Diagnostics
CAS Number:  
BCYE Agar/BCYE without Cysteine, Hardy Diagnostics
Catalog #: 76436-452
Supplier:  Hardy Diagnostics
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Format:
    Biplate
  • Packaging:
    Pack of 10
  • Description:
    BCYE agar/BCYE without cysteine
  • Size:
    15×100 mm
  • Cat. no.:
    76436-452
  • Fill Volume:
    12 ml/12 ml

Specifications

About this item

Buffered Charcoal Yeast Extract (BCYE) Agar formulations are recommended for use in the cultivation and primary isolation of Legionella spp. in water and other samples suspected of harboring the bacteria.

  • Optically clear polystyrene for distortion free viewing
  • Heavy-duty, reduces breakage during shipping
  • Polystyrene Petri plates feature raised rings for stacking ability and prevents sliding
  • Breathable packaging in sleeves of 10 plates in a 'breathable' bag that prevents buildup of condensation and excess moisture

Buffered Charcoal Yeast Extract (BCYE) Agar without Cysteine.

Customers Who Bought This Also Bought