Splash Guard, Material: Acrylic, Free-Standing, Color: Clear, with two legs to hold it, Eased and flame polished edges, 2-10inch x 12inch x 3/4inch legs, Dimension: 35x15x3/8inch
Catalog #: 76439-300
Supplier: Interdyne
Splash Guard, Material: Acrylic, Free-Standing, Color: Clear, with two legs to hold it, Eased and flame polished edges, 2-10inch x 12inch x 3/4inch legs, Dimension: 35x15x3/8inch
Catalog #: 76439-300
Supplier: Interdyne
Customers Who Bought This Also Bought
SP Bel-Art H-B® DURAC® Single-Channel Electronic Timer with 10-Button Direct Input and Certificate of Calibration, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
$48.82 to $442.63
VWR® Four-Channel Electronic Timer and Clock with Certificate of Calibration
Supplier: VWR International
$89.92 to $1,797.48
Shimpo LCD Contact/Non-Contact Handheld Laser Tachometers, Nidec Shimpo America
Supplier: Nidec Shimpo America
$771.89
Shimpo Compact LCD Contact and Contact/Non-Contact Tachometers, Nidec Shimpo America
Supplier: Nidec Shimpo America
$518.81
Shimpo Non-Contact Pocket Laser Tachometers, Nidec Shimpo America
Supplier: Nidec Shimpo America
$368.65