Splash Guard, Material: Acrylic, Color: Clear, Eased and flame polished edges, 45 deg Beveled Edge, Dimension: 25X18X0.50in
Catalog #: 76439-280
Supplier: Interdyne
Splash Guard, Material: Acrylic, Color: Clear, Eased and flame polished edges, 45 deg Beveled Edge, Dimension: 25X18X0.50in
Catalog #: 76439-280
Supplier: Interdyne
Customers Who Bought This Also Bought
VWR® Four-Channel Electronic Timer and Clock with Certificate of Calibration
Supplier: VWR International
$89.92 to $1,797.48
VWR® Water-Resistant/Shock-Resistant and Waterproof/Shockproof Stopwatches
Supplier: VWR International
$69.94