ContactUsLinkComponent
Package for first start up of the MD8 Airscan triclamp version, consists of the command unit, the stainless steel sampling head triclamp 50.5 version, 3m quick connector cable, stainless steel adapter triclamp and filter holder for the gelatin membrane filters
Catalog #: CA76409-218
Supplier:  Sartorius
Package for first start up of the MD8 Airscan triclamp version, consists of the command unit, the stainless steel sampling head triclamp 50.5 version, 3m quick connector cable, stainless steel adapter triclamp and filter holder for the gelatin membrane filters
Catalog #: CA76409-218
Supplier:  Sartorius
Customers Who Bought This Also Bought