ContactUsLinkComponent
Sampling Head, Triclamp 50.5 version for easy integration in tubes, part of the MD8 Airscan and actively creates a vacuum by sucking the air onto the GMF, and a quick connector cable (69898530) is needed, Line Voltage: 100 - 240 V +/- 10%, Dimensions: 128 mm x 112 mm x 154 mm
Catalog #: CA76409-214
Supplier:  Sartorius
Sampling Head, Triclamp 50.5 version for easy integration in tubes, part of the MD8 Airscan and actively creates a vacuum by sucking the air onto the GMF, and a quick connector cable (69898530) is needed, Line Voltage: 100 - 240 V +/- 10%, Dimensions: 128 mm x 112 mm x 154 mm
Catalog #: CA76409-214
Supplier:  Sartorius
Customers Who Bought This Also Bought