ContactUsLinkComponent
Command unit, for active air monitoring with the MD8 Airscan, operates the air sampler, allows the intake and display of data and stores up to 300 measurements, Air Sampling Method: Filtration, Stationary, quick connector cable, Voltage: 100-240 V, Dimensions: 282x102x107 mm
Catalog #: CA76409-208
Supplier:  Sartorius
Command unit, for active air monitoring with the MD8 Airscan, operates the air sampler, allows the intake and display of data and stores up to 300 measurements, Air Sampling Method: Filtration, Stationary, quick connector cable, Voltage: 100-240 V, Dimensions: 282x102x107 mm
Catalog #: CA76409-208
Supplier:  Sartorius
Customers Who Bought This Also Bought