Order Entry
ContactUsLinkComponent
Ultarfiltration unit, Vivaspin 20, Polyethersulfone, Molecular weight cut off: 3000, ultrafiltration unit with a unique twin membrane, Desalting and buffer exchange, suitable for starting volumes up to 20 ml for sample concentration with high process flexibility, Volume: 5-20 ml
Vivaspin® 20 Centrifugal Filtration Units, Sartorius
Catalog #: CA76409-098
Supplier:  Sartorius
Ultarfiltration unit, Vivaspin 20, Polyethersulfone, Molecular weight cut off: 3000, ultrafiltration unit with a unique twin membrane, Desalting and buffer exchange, suitable for starting volumes up to 20 ml for sample concentration with high process flexibility, Volume: 5-20 ml
Vivaspin® 20 Centrifugal Filtration Units, Sartorius
Catalog #: CA76409-098
Supplier:  Sartorius

Specifications

  • Membrane Material:
    Polyethersulfone (PES)
  • Max. RCF:
    8000 ×g
  • Ø×H:
    30×116 mm
  • Dead stop volume:
    50 µl
  • Hold-up volume:
    <20 µl
  • Filtration area:
    6.0 cm²
  • MWCO:
    3 kD

Specifications

About this item

Vivaspin® 20 centrifugal concentrators offer increased volume flexibility and performance to the Vivaspin® potfolio. Vivaspin® 20 handles 5 to 20 ml in swing bucket rotors and 14 ml in fixed angle rotors accepting 50 ml centrifuge tubes. Vivaspin® 20 features twin vertical membranes for unparalleled filtration speed and >100-fold concentration. Volume graduation allows monitoring the concentration process and the modified dead stop pocket simplifies pipetting of the final concentrate.

  • High retentate recovery >90%
  • Vertical twin membrane for ultra-fast concentration
  • Safe sample concentration due to dead-stop volume
  • Convenient concentrate recovery by pipetting
  • Volume graduation for easy reading/monitoring
  • Polycarbonate body and filterate vessel, polypropylene concentrator cap with acetal or aluminum pressure head

For higher process flexibility, it is available with unique accessories and operating methods. For one-step desalting following concentration, a diafiltration cup filled with buffer can be placed on the concentrator and spun another time to achieve 98% salt removal, instead of two spins to achieve the same result with the traditional refill and re-spin procedure.

When an appropriate centrifuge is unavailable, or for single sample processing, for gas pressure filtration, it can be filled with up to 15 ml and then pressurised for bench top concentration. For even faster processing, gas pressure filtration can be combined with centrifugation. 'Pressure-fugation' is particularly useful for difficult or viscous samples such as serum, when using a low process temperature, or generally when minimum process time is essential.

Customers Who Bought This Also Bought