Sartorius Vivaspin® Turbo 15
Catalog #: CA76409-062
Supplier: Sartorius
Specifications
- Membrane Material:Polyethersulfone (PES)
- Description:Vivaspin® Turbo 15
- Max. RCF:4000 ×g
- Ø×H:27×118/77 mm
- Dead stop volume:Swing bucket rotor: 100 µl; fixed angle rotor: 60 µl
- Hold-up volume:<10 µl
- Filtration area:7.2 cm²
- Molecular Weight:3K (cut-off)
Specifications
About this item
The Vivaspin® Turbo 15 can handle up to 15 ml sample volume in swing bucket rotors and 11 ml in fixed angle rotors accepting 50 ml centrifuge tubes.
- High chemical resistance
- High retentate recovery: more than 95%
- Dead stop volume: 60
- High flux PES membrane
- Universal rotor compatibility
The UV joining technology creates a smooth joint transition between membrane and plastic housing, which allows the collection of the complete concentrated sample into the pipette-friendly dead stop pocket.
The unit features a polystyrene butadiene copolymer body and a polyethersulfone membrane with a polystyrene cap.
The Vivaspin® Turbo 15’s sleek internal profile ensures maximum process speeds down to the last few microliters after 100-fold concentration.
Customers Who Bought This Also Bought