ContactUsLinkComponent
Purexall 10 Disinfectant
Disinfectant Spray-X, Innova chemicals
Catalog #: CA76423-586
Purexall 10 Disinfectant
Disinfectant Spray-X, Innova chemicals
Catalog #: CA76423-586
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Spray-X.
  • Size:
    946 mL
  • Cat. no.:
    CA76423-586
Specifications
About this item
Spray X ( Previously known as Purexall 10 ) is a ready-to-use, quaternary ammonium disinfectant spray designed as a general non-acid cleaner that is ideal for most environments including healthcare facilities, cannabis facilities, food processing facilities, schools, hotels and restaurants, kennels, veterinarians and animal hospitals, public transit, and retail locations due to its extensive list of disinfectant claims including tuberculosis, SARS associated Coronavirus and norovirus.
  • Included on the Health Canada Disinfectants for use against SARS-CoV-2 (COVID-19)
Spray X inhibits the growth of mold and mildew and controls related odors. It is Health Canada registered as a broad spectrum disinfectant cleaning product on hard, non-porous surfaces.
Caution: Use biocides safely. Always read the label and product information before use.
Customers Who Bought This Also Bought