ContactUsLinkComponent
Disinfectant Tablet
Easy Bleach™ Disinfectant Tablets, Berkshire
Catalog #: 76422-848
Supplier:  Berkshire
Disinfectant Tablet
Easy Bleach™ Disinfectant Tablets, Berkshire
Catalog #: 76422-848
Supplier:  Berkshire
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    EZ Bleach™ tablets
  • Size:
    1 tub of approx. 900 tablets (5g)
Specifications
About this item
NaDCC - Each rapidly dissolving 48.21% sodium dichloro-s-triazinetrione tablet is the equivalent of 29% available chlorine, a broad spectrum and sporicidal disinfectant.
  • No harsh fumes for the end user
  • Less corrosive to surfaces compared to liquid bleach
  • Neutral and consistent pH levels
  • Made in the USA and EPA registered
  • Kills C. difficile spores in 10 minutes, 99.9% effective against respiratory syncytial viruses and influenza H1N1 (colds and flu)
  • Flexibility – Easily control the level of dilution to match your specific disinfectant requirements
1 tub of approx. 900 tablets will produce 190 gallons of liquid bleach.
Tablets have a 3 year shelf life
Customers Who Bought This Also Bought