SP Wilmad-LabGlass Large Writing Area High-Throughput NMR Tubes, 5 mm O.D., SP Industries
Catalog #: 76420-860
Supplier: SP Industries
Specifications
- Length:178 mm (7")
- O.D.:4.94665±0.01905 mm
- Wall thickness:0.43 mm
- Description:High-throughput NMR tubes
Specifications
About this item
Double the writing area allows for easy sample marking and identification.
- 2x larger marking area eliminates cramped and unreadable sample names
- Silkscreened writing area keeps sample names from easily wiping off
- Designed for routine use in most low to mid field NMR spectrometers
- Camber and concentricity values listed are the Total Indicator Reading (TIR)
- Made from ASTM E438 Type 1 Class B glass
- 100% inspected for surface defects and physical dimensions
These High-Throughput tubes have an average camber of 60 µm to ensure spectrum clarity for small molecule (MW<250) samples up to 600 MHz.
Customers Who Bought This Also Bought