Dual Ended Stainless Steel Micro Lab Spatula, Eisco
Catalog #: 470328-218
Supplier: Wards
Specifications
- Length:7.9"
- Description:Dual ended spatula
- Material:Stainless steel
- Cat. no.:470328-218
Specifications
About this item
Features one tapered spatula end and one rounded flat spatula end. Designed for use with small samples. Perfect for a variety of laboratory uses.
- Made of high quality polished stainless steel
- Autoclavable, reusable
- Feature one tapered spatula end and one rounded spatula end
- Designed for use with small samples
Stainless steel lab scoop with a tapered flat spatula on one end and rounded flat spatula on the other end. Total legnth is 7.9". Great for adding small amounts of ingredients and mixing.
Customers Who Bought This Also Bought