Teflon Coated Stainless Steel Lab Scoop with Spatula, Eisco
Catalog #: 470328-206
Supplier: Wards
Specifications
- Length:5.9"
- Description:Lab scoop with spatula
- Material:Teflon® coated
Specifications
About this item
Features one scoop end and one spatula end with knife edge. Perfect for a variety of laboratory uses.
- Made of high quality stainless steel
- Coated in non-stick Teflon
- Extremely resistant to chemicals
- Autoclavable, reusable
- Feature one scoop end and one spatula end
Teflon coated stainless steel lab scoop with spoon on one end and flat edge with on the other end. Total length 5.9". These scoops are great for adding small amounts of ingredients and mixing.
Customers Who Bought This Also Bought