VWR® Screw Cap Microtubes
Catalog #: 76417-242
Supplier: VWR International
Specifications
- Format:Conical bottom
- Sterility:Non sterile
- Volume:1.5 ml
- Color:Clear
- Description:Microtubes without ribbing
- Maximum RCF:20000×g
- Graduations:Yes
Specifications
About this item
VWR® screw cap microtubes are the perfect environment to securely store your important samples and reagents.
- All tubes are manufactured from ultra-pure PP
- High-clarity tubes provide excellent visibility to the sample
- O-ringed flat caps feature a ³/₄ turn to open or close
- Wide temperature range to meet a wide variety of applications: −196 to +121 °C, but not suitable for immersion in liquid nitrogen
- Sterile assemblies or handy, non sterile combo packs are available
Screw cap microtubes are available in the three most commonly volumes and available with conical or free-standing/skirted bottoms.
Certifications: Certified free of detectable DNase, RNase, DNA, PCR inhibitors. Sterile items are also free of pyrogens. The PP used in tubes meets FDA 21 CFR 177.1520 & USP Class VI standards.
Customers Who Bought This Also Bought