VWR® Flat Caps for Screw Cap Microtubes
Catalog #: 76417-220
Supplier: VWR International
Specifications
- Sterility:Non sterile
- Cat. no.:76417-220
- Cap Color:Opaque green
Specifications
About this item
Flat caps with O-ring for use with VWR® screw cap microtubes only (series 76417-xxx).
Feature ³/₄ turn to easily open and close tubes requiring less revolutions than competitor tubes.
Customers Who Bought This Also Bought