North America power cable (NEMA 5-15) for AquaMate 7100 & 8100 Spectrophotometers
Catalog #: 76415-514
Supplier: Thermo Fisher Scientific
North America power cable (NEMA 5-15) for AquaMate 7100 & 8100 Spectrophotometers
Catalog #: 76415-514
Supplier: Thermo Fisher Scientific
Customers Who Bought This Also Bought
Ethyl acetate ≥99.8%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Supplier: VWR International
458,88 $ to 1 237,78 $
Formalin 10%, VWR®, Reagent Grade for histology, neutral buffered
Supplier: VWR International
139,70 $
Sodium hypochlorite 5% available chlorine in aqueous solution, BAKER ANALYZED®, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
195,79 $ to 1 600,59 $