Sample holder, assembled in a removable sample compartment liner, for film/filter for Aquamate 7100/8100 Spectrophotometer, Holds Filters/lenses up to 50mm long, 80mm tall and 6mm thick, Includes: 1 Film / Filter Holder
Catalog #: 76415-504
Supplier: Thermo Fisher Scientific
Sample holder, assembled in a removable sample compartment liner, for film/filter for Aquamate 7100/8100 Spectrophotometer, Holds Filters/lenses up to 50mm long, 80mm tall and 6mm thick, Includes: 1 Film / Filter Holder
Catalog #: 76415-504
Supplier: Thermo Fisher Scientific
Customers Who Bought This Also Bought
Sodium hydroxide 40% (w/w) in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
146,85 $ to 576,51 $
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
684,63 $ to 2 738,51 $
Sol-Vet® Standard Insulin Syringes (Veterinary Care), Sol M
Supplier: SOL-MILLENNIUM MEDICAL
27,21 $ to 266,64 $
Sodium chloride ≥99.0% (by argentometric titration), crystals, AR® ACS, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
116,87 $ to 467,59 $
tri-Sodium citrate dihydrate ≥99% (by anhydrous basis), granular BP, Ph. Eur., JP, USP, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
3 278,39 $