Order Entry
ContactUsLinkComponent
VWR* Hardness test strip, Quick and easy determination of water hardness, for 5-21 tests, convenient dip and read procedure and the clear colour changes from green to red ensure reliable results within seconds, plays important role, Gradation: 90 ->180 ->270->360 ->445 ppm, 500 dosatest
Test Strips, Water Hardness, Dosatest, VWR Chemicals BDH®
Catalog #: BDH85442.601
Supplier:  VWR International
CAS Number:  
VWR* Hardness test strip, Quick and easy determination of water hardness, for 5-21 tests, convenient dip and read procedure and the clear colour changes from green to red ensure reliable results within seconds, plays important role, Gradation: <55 ->90 ->180 ->270->360 ->445 ppm, 500 dosatest
Test Strips, Water Hardness, Dosatest, VWR Chemicals BDH®
Catalog #: BDH85442.601
Supplier:  VWR International
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Hardness 5 - 25 °d
  • Range:
    0 ·  5 ·10 · 15 ·20 · 25
  • No. of Strips:
    100

Specifications

About this item

A ready-to-use kit that fits into your lab coat pocket.

  • Rapid and convenient
  • Easy to handle
  • Choice of ranges for required precision

Ideal to monitor water hardness, especially suitable for boiler testing, aquaria and laundries. Semi-quantitative results can be obtained between 10 and 120 seconds.

The color charts are adjusted and checked using certified standard solutions that are directly traceable to primary NIST standards.

1 mmol/litre =10 °f = 5.60 °d= 7,022 °e

1 mmol/litre= 100.09 mg/l CaCO₃

Packaging: Container with 100 test strips 6 x 95 mm
Customers Who Bought This Also Bought